ENA341295834 Enamine

4-amino-1-(2-methylcyclopropyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD20502139
  • Cas Number : 1354951-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94273
Availability details for
Quantity
  • Availability
  • Add to Cart