ENA341295835 Enamine

3-methyl-2-phenylazetidine

  • MDL number MFCD20502140
  • Cas Number : 1354958-21-6
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94281
Availability details for
Quantity
  • Availability
  • Add to Cart