ENA341295841 Enamine

2-(2-methyl-1H-imidazol-1-yl)pyridine-4-carboxylic acid hydrochloride

  • MDL number MFCD20502147
  • Cas Number : 1354957-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94386
Availability details for
Quantity
  • Availability
  • Add to Cart