ENA341295848 Enamine

2-{6-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl}acetic acid

  • MDL number MFCD11215327
  • Cas Number : 1160245-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92549
Availability details for
Quantity
  • Availability
  • Add to Cart