ENA341295853 Enamine

5-methyl-4-[(morpholin-4-yl)methyl]-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD06804405
  • Cas Number : 893750-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92554
Availability details for
Quantity
  • Availability
  • Add to Cart