ENA341295858 Enamine

3-cyclopropyl-2-ethyl-6-oxo-2H,6H,7H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD10699090
  • Cas Number : 1160246-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92559
Availability details for
Quantity
  • Availability
  • Add to Cart