ENA341295863 Enamine

3-methyl-1-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08700649
  • Cas Number : 113100-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92564
Availability details for
Quantity
  • Availability
  • Add to Cart