ENA341295872 Enamine

2-(3-carbamoyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD06805792
  • Cas Number : 1006458-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92573
Availability details for
Quantity
  • Availability
  • Add to Cart