ENA341295876 Enamine

2-{[4-(difluoromethyl)-2-methyl-2H-pyrazolo[3,4-b]pyridin-6-yl]sulfanyl}acetic acid

  • MDL number MFCD09859245
  • Cas Number : 1018166-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92577
Availability details for
Quantity
  • Availability
  • Add to Cart