ENA341295881 Enamine

3-cyclopropyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08696531
  • Cas Number : 1006486-60-7
  • Molecular Weight : 234.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92582
Availability details for
Quantity
  • Availability
  • Add to Cart