ENA341295894 Enamine

5-(4-methylphenoxymethyl)thiophene-2-carboxylic acid

  • MDL number MFCD06804715
  • Cas Number : 934155-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92595

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92595
Availability details for
Quantity
  • Availability
  • Add to Cart