ENA341295897 Enamine

3-amino-1-(butan-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06805286
  • Cas Number : 1006494-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92598
Availability details for
Quantity
  • Availability
  • Add to Cart