ENA341295898 Enamine

3-(3-methoxyphenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD09701747
  • Cas Number : 1018143-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92599
Availability details for
Quantity
  • Availability
  • Add to Cart