ENA341295907 Enamine

5-[(2,6-dichlorophenoxy)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06804730
  • Cas Number : 74556-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92608
Availability details for
Quantity
  • Availability
  • Add to Cart