ENA341295910 Enamine

5-(2-chloro-4-fluorophenoxymethyl)thiophene-2-carboxylic acid

  • MDL number MFCD06804713
  • Cas Number : 934127-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92611
Availability details for
Quantity
  • Availability
  • Add to Cart