ENA341295919 Enamine

5-[(4-amino-1H-pyrazol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD09701810
  • Cas Number : 1170539-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92620
Availability details for
Quantity
  • Availability
  • Add to Cart