ENA341295920 Enamine

5-(propan-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD08696379
  • Cas Number : 29481-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-82
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92621
Availability details for
Quantity
  • Availability
  • Add to Cart