ENA341295922 Enamine

1-ethyl-3-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06805247
  • Cas Number : 1006441-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92623
Availability details for
Quantity
  • Availability
  • Add to Cart