ENA341295925 Enamine

1-(1-carboxypropyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09701797
  • Cas Number : 1170851-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92626
Availability details for
Quantity
  • Availability
  • Add to Cart