ENA341295926 Enamine

1-ethyl-6-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06805326
  • Cas Number : 934062-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92627
Availability details for
Quantity
  • Availability
  • Add to Cart