ENA341295930 Enamine

2-{3-cyclopropyl-4,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-1-yl}acetic acid

  • MDL number MFCD08696715
  • Cas Number : 937605-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92631
Availability details for
Quantity
  • Availability
  • Add to Cart