ENA341295933 Enamine

3-[4-(methoxycarbonyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD09701618
  • Cas Number : 1170943-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92634
Availability details for
Quantity
  • Availability
  • Add to Cart