ENA341295936 Enamine

1-(1-methyl-1H-pyrazol-3-yl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09834170
  • Cas Number : 1171097-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92637
Availability details for
Quantity
  • Availability
  • Add to Cart