ENA341295938 Enamine

2-methyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD09701470
  • Cas Number : 1018050-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92639
Availability details for
Quantity
  • Availability
  • Add to Cart