ENA341295940 Enamine

5-[(2-methoxyphenoxy)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06804708
  • Cas Number : 934080-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92641
Availability details for
Quantity
  • Availability
  • Add to Cart