ENA341295949 Enamine

4-chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04968048
  • Cas Number : 1006489-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92650
Availability details for
Quantity
  • Availability
  • Add to Cart