ENA341295950 Enamine

3-{[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]carbamoyl}propanoic acid

  • MDL number MFCD03072831
  • Cas Number : 522622-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92651
Availability details for
Quantity
  • Availability
  • Add to Cart