ENA341295951 Enamine

2-[4-(2,2,2-trifluoroethoxy)phenyl]acetic acid

  • MDL number MFCD08696483
  • Cas Number : 170361-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92652
Availability details for
Quantity
  • Availability
  • Add to Cart