ENA341295954 Enamine

1-[(3-methylphenyl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06805380
  • Cas Number : 1006494-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92655
Availability details for
Quantity
  • Availability
  • Add to Cart