ENA341295959 Enamine

2-(3-amino-5-methyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD04969592
  • Cas Number : 1006455-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92660
Availability details for
Quantity
  • Availability
  • Add to Cart