ENA341295960 Enamine

1-[(3-chlorophenyl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06805384
  • Cas Number : 1006494-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92661

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92661
Availability details for
Quantity
  • Availability
  • Add to Cart