ENA341295962 Enamine

2-(4-amino-5-methyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD09701838
  • Cas Number : 1172349-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92663
Availability details for
Quantity
  • Availability
  • Add to Cart