ENA341295966 Enamine

3-(4-chloro-1-methyl-1H-pyrazol-3-yl)propanoic acid

  • MDL number MFCD06808505
  • Cas Number : 1006483-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92667
Availability details for
Quantity
  • Availability
  • Add to Cart