ENA341295968 Enamine

3-(1,5-dimethyl-1H-pyrazol-4-yl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD06805567
  • Cas Number : 1006496-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92669
Availability details for
Quantity
  • Availability
  • Add to Cart