ENA341295969 Enamine

1-[(1-ethyl-5-methyl-1H-pyrazol-4-yl)methyl]piperidine-4-carboxylic acid

  • MDL number MFCD06805662
  • Cas Number : 1006483-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92670
Availability details for
Quantity
  • Availability
  • Add to Cart