ENA341295975 Enamine

4-bromo-1-cyclopentyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805231
  • Cas Number : 1006451-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92676
Availability details for
Quantity
  • Availability
  • Add to Cart