ENA341295976 Enamine

3-amino-4-chloro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD01528031
  • Cas Number : 351990-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92677
Availability details for
Quantity
  • Availability
  • Add to Cart