ENA341295977 Enamine

2-[4-chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD04969232
  • Cas Number : 1006446-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92678
Availability details for
Quantity
  • Availability
  • Add to Cart