ENA341295984 Enamine

4-bromo-1-cyclopentyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06805245
  • Cas Number : 1006494-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92685
Availability details for
Quantity
  • Availability
  • Add to Cart