ENA341295986 Enamine

2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

  • MDL number MFCD04967809
  • Cas Number : 1006348-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92687
Availability details for
Quantity
  • Availability
  • Add to Cart