ENA341295989 Enamine

3-chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidine-7-carboxylic acid

  • MDL number MFCD08556254
  • Cas Number : 932236-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92690
Availability details for
Quantity
  • Availability
  • Add to Cart