ENA341295993 Enamine

1-[(trimethyl-1H-pyrazol-4-yl)methyl]piperidine-3-carboxylic acid

  • MDL number MFCD06805667
  • Cas Number : 1006457-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92694
Availability details for
Quantity
  • Availability
  • Add to Cart