ENA341295997 Enamine

3-cyclopropyl-1-ethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04970942
  • Cas Number : 1006446-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92698
Availability details for
Quantity
  • Availability
  • Add to Cart