ENA341296001 Enamine

1-(2,2-difluoroethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08696527
  • Cas Number : 1006462-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92702
Availability details for
Quantity
  • Availability
  • Add to Cart