ENA341296011 Enamine

4-[(4-chlorophenoxy)methyl]-5-methyl-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD08685648
  • Cas Number : 905809-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92712
Availability details for
Quantity
  • Availability
  • Add to Cart