ENA341296014 Enamine

5-chloro-6-cyclopropyl-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09473344
  • Cas Number : 1011396-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92715
Availability details for
Quantity
  • Availability
  • Add to Cart