ENA341296018 Enamine

5-[(2,4-difluorophenoxy)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06804716
  • Cas Number : 934127-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92719
Availability details for
Quantity
  • Availability
  • Add to Cart