ENA341296026 Enamine

1-[(4-chloro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967760
  • Cas Number : 1006473-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92727
Availability details for
Quantity
  • Availability
  • Add to Cart