ENA341296029 Enamine

4-chloro-1-[(1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967772
  • Cas Number : 1006473-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92730
Availability details for
Quantity
  • Availability
  • Add to Cart