ENA341296030 Enamine

2-[4-(difluoromethyl)-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

  • MDL number MFCD08697720
  • Cas Number : 937607-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92731
Availability details for
Quantity
  • Availability
  • Add to Cart